Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320586.1 | NP_001307515.1 | 6 - 7 | 662 | 80 | |
| NM_001320587.1 | NP_001307516.1 | 5 - 6 | 569 | 80 | ||
| NM_001320588.1 | NP_001307517.1 | 3 - 4 | 363 | 80 | ||
| NM_138448.3 | NP_612457.1 | 3 - 4 | 614 | 80 | ||
| XM_017005411.1 | XP_016860900.1 | 7 - 8 | 1008 | 80 | ||
| GenBank mRNA | BC012290.1 | - | 3 - 4 | 383 | 3864319 | 80 |
| BF671963.1 | - | 3 - 4 | 362 | 80 | ||
| BG741807.1 | - | 5 - 6 | 563 | 80 | ||
| BU543943.1 | - | 6 - 7 | 662 | 80 | ||
| X84195.1 | - | 3 - 4 | 363 | 80 | ||
More Information
Set Membership: |
|

ACYP2
Genomic Map