Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC019342.2 | - | 6 - 7 | 713 | 3357734 | 102 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPAN18| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC019342.2 | - | 6 - 7 | 713 | 3357734 | 102 |
Set Membership: |
|