Assay Details
| Gene Symbol: MAPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123066.3 | NP_001116538.2 | 10 - 11 | 1879 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000592.hg.1, TC17_ctg5_hap1000017.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
neuron migration
brain development
female pregnancy
response to nutrient
adult walking behavior
axo-dendritic transport
intrinsic apoptotic signaling pathway in response to oxidative stress
response to organic substance
regulation of autophagy
regulation of microtubule polymerization
positive regulation of microtubule polymerization
negative regulation of intracellular transport
positive regulation of axon extension
mitochondrion transport along microtubule
axon extension
generation of neurons
regulation of microtubule-based movement
structural constituent of cytoskeleton
protein binding
microtubule binding
SH3 domain binding
enzyme binding
protein kinase binding
protein complex binding
apolipoprotein binding
protein phosphatase 2A binding
lipoprotein particle binding
microtubule cytoskeleton organization
neuron migration
brain development
female pregnancy
response to nutrient
adult walking behavior
axo-dendritic transport
intrinsic apoptotic signaling pathway in response to oxidative stress
response to organic substance
regulation of autophagy
regulation of microtubule polymerization
positive regulation of microtubule polymerization
negative regulation of intracellular transport
positive regulation of axon extension
mitochondrion transport along microtubule
axon extension
generation of neurons
regulation of microtubule-based movement
structural constituent of cytoskeleton
protein binding
microtubule binding
SH3 domain binding
enzyme binding
protein kinase binding
protein complex binding
apolipoprotein binding
protein phosphatase 2A binding
lipoprotein particle binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000262410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000334239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000340799.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000344290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000347967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000351559.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000354326.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000415613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000420682.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000431008.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000446361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | ENST00000535772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_001123066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_001123067.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_001203251.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_001203252.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_005910.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_016834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_016835.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | NM_016841.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007260.hg.1 | TC17000592.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | TC17_ctg5_hap1000017.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_001123066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_001123067.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_001203251.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_001203252.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_005910.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_016834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_016835.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000217.hg.1 | NM_016841.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000262410.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000334239.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000340799.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000344290.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000351559.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000415613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000420682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000431008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000446361.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000535772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000570299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000571987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000574436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000576518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_001123066.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_001203252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_016834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_016835.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_016841.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000262410.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000334239.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000340799.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000344290.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000351559.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000415613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000420682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000431008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000446361.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000535772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000570299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000571987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000574436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | ENST00000576518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_001123066.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_001203252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_016834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_016835.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | NM_016841.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159258.hg.1 | TC1700008082.hg.1 | Clariom D TC |

MAPT
Genomic Map