Assay Details
| Gene Symbol: PIK3C2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321378.1 | NP_001308307.1 | 12 - 13 | 2383 | 136 | |
| NM_001321380.1 | NP_001308309.1 | 10 - 11 | 1184 | 136 | ||
| NM_002645.3 | NP_002636.2 | 11 - 12 | 2314 | 136 | ||
| XM_011520186.2 | XP_011518488.1 | 11 - 12 | 2309 | 136 | ||
| XM_017017924.1 | XP_016873413.1 | 12 - 13 | 2330 | 136 | ||
| GenBank mRNA | AK294416.1 | - | 10 - 11 | 1171 | 136 | |
| BC113658.1 | - | 10 - 11 | 2166 | 8322710 | 136 | |
| BX648778.1 | - | 1 - 2 | 250 | 136 | ||
| Y13367.1 | - | 10 - 11 | 2100 | 136 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003476.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
exocytosis
endocytosis
epidermal growth factor receptor signaling pathway
insulin receptor signaling pathway
vascular smooth muscle contraction
phosphatidylinositol-3-phosphate biosynthetic process
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
clathrin coat assembly
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
phosphatidylinositol binding
phosphatidylinositol biosynthetic process
exocytosis
endocytosis
epidermal growth factor receptor signaling pathway
insulin receptor signaling pathway
vascular smooth muscle contraction
phosphatidylinositol-3-phosphate biosynthetic process
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
clathrin coat assembly
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
phosphatidylinositol binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11018279.hg.1 | TC11003476.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018279.hg.1 | ENST00000265970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018279.hg.1 | ENST00000540361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018279.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203729.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203729.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203729.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203729.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203729.hg.1 | TC1100013148.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |

PIK3C2A
Genomic Map