Assay Details
| Gene Symbol: PIK3C2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321378.1 | NP_001308307.1 | 9 - 10 | 1991 | 106 | |
| NM_001321380.1 | NP_001308309.1 | 7 - 8 | 792 | 106 | ||
| NM_002645.3 | NP_002636.2 | 8 - 9 | 1922 | 106 | ||
| XM_011520186.2 | XP_011518488.1 | 8 - 9 | 1917 | 106 | ||
| XM_017017924.1 | XP_016873413.1 | 9 - 10 | 1938 | 106 | ||
| GenBank mRNA | AK294416.1 | - | 7 - 8 | 779 | 106 | |
| AK299474.1 | - | 1854 | 106 | |||
| BC113658.1 | - | 7 - 8 | 1774 | 8322710 | 106 | |
| Y13367.1 | - | 7 - 8 | 1708 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003476.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
exocytosis
endocytosis
epidermal growth factor receptor signaling pathway
insulin receptor signaling pathway
vascular smooth muscle contraction
phosphatidylinositol-3-phosphate biosynthetic process
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
clathrin coat assembly
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
phosphatidylinositol binding
phosphatidylinositol biosynthetic process
exocytosis
endocytosis
epidermal growth factor receptor signaling pathway
insulin receptor signaling pathway
vascular smooth muscle contraction
phosphatidylinositol-3-phosphate biosynthetic process
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
clathrin coat assembly
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
phosphatidylinositol binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11018282.hg.1 | TC11003476.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018282.hg.1 | ENST00000265970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018282.hg.1 | ENST00000540361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018282.hg.1 | ENST00000544896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018282.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018283.hg.1 | TC11003476.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018283.hg.1 | ENST00000265970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018283.hg.1 | ENST00000540361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018283.hg.1 | ENST00000544896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018283.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203732.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203732.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203732.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203732.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203732.hg.1 | TC1100013148.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000265970.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000503729.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000532035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | ENST00000533645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013148.hg.1 | NM_002645.3 | Refseq |

PIK3C2A
Genomic Map