Assay Details
| Gene Symbol: KCNU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134267.1 | 24 - 25 | 2824 | 61 | ||
| GenBank mRNA | BC028701.2 | - | 24 - 25 | 2765 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000518904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000518904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000399881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000518904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000518904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000522417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | ENST00000523973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007284.hg.1 | NM_001031836.2 | Refseq |

KCNU1
Genomic Map