Assay Details
| Gene Symbol: KCNU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134267.1 | 25 - 26 | 2882 | 75 | ||
| GenBank mRNA | BC028701.2 | - | 25 - 26 | 2823 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

KCNU1
Genomic Map