Assay Details
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113434.4 | NP_001106905.1 | 2 | 4292 | 98 | |
GenBank mRNA | AK123143.1 | - | 1 | 667 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001266.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17016959.hg.1 | ENST00000391411.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17016959.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17016959.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17016959.hg.1 | TC17001266.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17016956.hg.1 | ENST00000391411.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17016956.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17016956.hg.1 | TC17001266.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17016958.hg.1 | ENST00000391411.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17016958.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17016958.hg.1 | TC17001266.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700179231.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179231.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700179231.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179231.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700179231.hg.1 | TC1700010101.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700179234.hg.1 | TC1700010101.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700179233.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179233.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700179233.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700179233.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700179233.hg.1 | TC1700010101.hg.1 | Clariom D TC |
