Assay Details
| Gene Symbol: MUC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF125525.1 | - | 1 - 2 | 54 | 102 | |
| EF670706.1 | - | 1 - 2 | 104 | 102 | ||
| EF670707.1 | - | 1 - 2 | 104 | 102 | ||
| EF670715.1 | - | 1 - 2 | 104 | 102 | ||
| Z17324.1 | - | 1 - 2 | 127 | 102 | ||
More Information
Set Membership: |
|

MUC1
Genomic Map