Assay Details
Gene Symbol: OTUD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC020622.1 | - | 1 - 2 | 84 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC020622.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000417191.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000581312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | ENST00000619871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015717.hg.1 | NM_020205.3 | Refseq |
