Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017316.hg.1 |
ENST00000005905.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017316.hg.1 |
ENST00000389003.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017316.hg.1 |
ENST00000528896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017316.hg.1 |
ENST00000544884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017316.hg.1 |
NM_014680.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017316.hg.1 |
TC17001302.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
ENST00000389003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
ENST00000528896.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
ENST00000544884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
NM_014680.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
ENST00000389003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
ENST00000528896.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
ENST00000544884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
NM_014680.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700179997.hg.1 |
TC1700010202.hg.1 |
Clariom D TC |