Assay Details
Gene Symbol: MELTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033316.3 | NP_201573.1 | 6 - 7 | 820 | 61 | |
GenBank mRNA | BC001875.1 | - | 6 - 7 | 789 | 61 | |
BC002623.2 | - | 6 - 7 | 775 | 61 | ||
BC007550.1 | - | 6 - 7 | 789 | 61 | ||
BC071910.1 | - | 6 - 7 | 780 | 61 | ||
BC093920.1 | - | 6 - 7 | 770 | 61 | ||
BC111947.1 | - | 6 - 7 | 770 | 61 | ||
BC143257.1 | - | 6 - 7 | 689 | 61 | ||
BT007377.1 | - | 6 - 7 | 707 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03002173.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
positive regulation of plasminogen activation
regulation of cell proliferation
iron ion homeostasis
positive regulation of extracellular matrix disassembly
iron ion import
negative regulation of substrate adhesion-dependent cell spreading
iron ion binding
protein binding
regulation of cell growth
positive regulation of plasminogen activation
regulation of cell proliferation
iron ion homeostasis
positive regulation of extracellular matrix disassembly
iron ion import
negative regulation of substrate adhesion-dependent cell spreading
iron ion binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03038374.hg.1 | TC03002173.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03038374.hg.1 | ENST00000296351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03038374.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000469783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000469783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000469783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000489445.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000489445.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000489445.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000491399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000491399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | ENST00000491399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202628.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202628.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202628.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202628.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202628.hg.1 | TC0300013733.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300202629.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202629.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202629.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202629.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202629.hg.1 | TC0300013733.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296350.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000296351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000439320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000469783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000469783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000469783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000473501.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000489445.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000489445.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000489445.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000491399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000491399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | ENST00000491399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_005929.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013733.hg.1 | NM_033316.3 | Refseq |
