Assay Details
| Gene Symbol: KAT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006721818.3 | XP_006721881.1 | 6 - 7 | 1042 | 77 | |
| XM_017024448.1 | XP_016879937.1 | 6 - 7 | 1293 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006721818.3, XM_017024448.1 transcripts.
Set Membership: |
|

KAT2A
Genomic Map