Assay Details
Gene Symbol: ZNF581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016535.3 | NP_057619.1 | 1 - 2 | 153 | 67 | |
GenBank mRNA | AF151023.2 | - | 1 - 2 | 153 | 67 | |
AK026203.1 | - | 1 - 2 | 153 | 67 | ||
AK315603.1 | - | 1 - 2 | 124 | 67 | ||
BC001219.1 | - | 1 - 2 | 122 | 3528677 | 67 | |
BC010097.2 | - | 1 - 2 | 125 | 3504825 | 67 | |
BC071845.1 | - | 1 - 2 | 107 | 4764567 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000903.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF151023.2,AK315603.1,BC001219.1,BC010097.2,BC071845.1,NM_016535.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19012507.hg.1 | ENST00000270451.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19012507.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19012507.hg.1 | TC19000903.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19012512.hg.1 | ENST00000270451.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19012512.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19012512.hg.1 | TC19000903.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000585995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000585995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000592461.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000592461.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000592461.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19012510.hg.1 | ENST00000270451.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19012510.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19012510.hg.1 | TC19000903.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000585995.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | TC1900011804.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000585995.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202755.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000325333.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000543039.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000545125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000585995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000585995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000587252.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000588537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000590190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000592461.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000592461.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000592461.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | ENST00000592881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_001163423.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016202.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011804.hg.1 | NM_207115.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900202749.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202749.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900202749.hg.1 | TC1900011804.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900202749.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202749.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900202752.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202752.hg.1 | NM_016535.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900202752.hg.1 | TC1900011804.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900202752.hg.1 | ENST00000270451.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202752.hg.1 | NM_016535.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 201589 | - | - |
