Assay Details
| Gene Symbol: ADCY8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115.2 | NP_001106.1 | 9 - 10 | 2469 | 109 | |
| XM_005250769.3 | XP_005250826.1 | 9 - 10 | 2592 | 109 | ||
| XM_006716501.3 | XP_006716564.1 | 8 - 9 | 2598 | 109 | ||
| XM_017013006.1 | XP_016868495.1 | 8 - 9 | 2360 | 109 | ||
| GenBank mRNA | GQ472223.1 | - | 9 - 10 | 4307 | 109 | |
| M83533.1 | - | 3 - 4 | 485 | 109 | ||
| Z35309.1 | - | 9 - 10 | 4307 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001646.hg.1
Set Membership: |
|

ADCY8
Genomic Map