Assay Details
| Gene Symbol: ENPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040092.2 | NP_001035181.1 | 11 - 12 | 1057 | 82 | |
| NM_001130863.2 | NP_001124335.1 | 11 - 12 | 1057 | 82 | ||
| XM_017013571.1 | XP_016869060.1 | 11 - 12 | 1091 | 82 | ||
| XM_017013572.1 | XP_016869061.1 | 11 - 12 | 1035 | 82 | ||
| XM_017013573.1 | XP_016869062.1 | 11 - 12 | 1091 | 82 | ||
| XM_017013574.1 | XP_016869063.1 | 11 - 12 | 1091 | 82 | ||
| XM_017013575.1 | XP_016869064.1 | 11 - 12 | 1035 | 82 | ||
| GenBank mRNA | AK315253.1 | - | 11 - 12 | 1030 | 82 | |
| BC034961.2 | - | 11 - 12 | 1030 | 4828484 | 82 | |
| BQ365938.1 | - | 2 - 3 | 82 | |||
| D45421.1 | - | 11 - 12 | 1004 | 82 | ||
| EU131011.2 | - | 11 - 12 | 971 | 82 | ||
More Information
Set Membership: |
|

ENPP2
Genomic Map