Assay Details
| Gene Symbol: SVOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018711.4 | NP_061181.1 | 6 - 7 | 950 | 57 | |
| GenBank mRNA | BC033587.1 | - | 6 - 7 | 750 | 5243017 | 57 |
| BC094722.1 | - | 6 - 7 | 597 | 4817829 | 57 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001943.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12025297.hg.1 | ENST00000299134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025297.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025297.hg.1 | TC12001943.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191945.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191945.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191945.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191945.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191945.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191945.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191945.hg.1 | TC1200011855.hg.1 | Clariom D TC |

SVOP
Genomic Map