Assay Details
| Gene Symbol: CDH26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021810.4 | NP_068582.2 | 1 - 2 | 224 | 96 | |
| GenBank mRNA | AK074477.1 | - | 1 - 2 | 212 | 96 | |
| BC062570.1 | - | 1 - 2 | 224 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000474.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK074477.1,BC062570.1,NM_021810.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20006611.hg.1 | ENST00000244049.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006611.hg.1 | ENST00000350849.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006611.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006611.hg.1 | TC20000474.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000348616.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000370991.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000477058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | ENST00000497614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007943.hg.1 | NM_177980.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | NM_021810.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | TC2000007943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | ENST00000244049.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | ENST00000350849.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | ENST00000456106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155271.hg.1 | NM_021810.5 | Refseq |

CDH26
Genomic Map