Assay Details
Gene Symbol: FAM110A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031424.5 | NP_113612.1 | 1 | 43 | 54 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000012.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_031424.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20000124.hg.1 | ENST00000541082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000124.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000124.hg.1 | TC20000012.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144274.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144274.hg.1 | TC2000006464.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144274.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144272.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144272.hg.1 | TC2000006464.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144272.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000246100.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000304189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000381941.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000505691.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | ENST00000541082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001042353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289145.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289146.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_001289147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_031424.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_207121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006464.hg.1 | NM_207121.4 | Refseq |
