Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008497.hg.1 |
TC07000582.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008497.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008497.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158889.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158889.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158889.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158889.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158889.hg.1 |
TC0700008450.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000297293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
ENST00000493372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008450.hg.1 |
NM_014916.3 |
Refseq |