Assay Details
| Gene Symbol: LMTK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014916.3 | NP_055731.2 | 1 - 2 | 396 | 112 | |
| GenBank mRNA | AB029002.1 | - | 1 - 2 | 396 | 112 | |
| AY130988.1 | - | 1 - 2 | 103 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000582.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
receptor recycling
protein phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
endocytic recycling
transferrin transport
negative regulation of catalytic activity
early endosome to late endosome transport
protein autophosphorylation
protein serine/threonine kinase activity
protein phosphatase inhibitor activity
protein binding
ATP binding
myosin VI binding
receptor recycling
protein phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
endocytic recycling
transferrin transport
negative regulation of catalytic activity
early endosome to late endosome transport
protein autophosphorylation
protein serine/threonine kinase activity
protein phosphatase inhibitor activity
protein binding
ATP binding
myosin VI binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07008487.hg.1 | TC07000582.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008487.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008487.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008487.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008484.hg.1 | TC07000582.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008484.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008484.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008485.hg.1 | TC07000582.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008485.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008485.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158877.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158877.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158877.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158877.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158877.hg.1 | TC0700008450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700158879.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158879.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158879.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158879.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158879.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158879.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158879.hg.1 | TC0700008450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700158876.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158876.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158876.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158876.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158876.hg.1 | TC0700008450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000297293.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | ENST00000493372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008450.hg.1 | NM_014916.3 | Refseq |

LMTK2
Genomic Map