Assay Details
| Gene Symbol: GOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286220.1 | NP_001273149.1 | 5 - 6 | 696 | 66 | |
| NM_002080.3 | NP_002071.2 | 6 - 7 | 825 | 66 | ||
| GenBank mRNA | AK223271.1 | - | 6 - 7 | 760 | 66 | |
| AK290847.1 | - | 6 - 7 | 758 | 66 | ||
| AK295993.1 | - | 5 - 6 | 665 | 66 | ||
| AK302249.1 | - | 5 - 6 | 666 | 66 | ||
| AK307598.1 | - | 6 - 7 | 712 | 66 | ||
| BC000525.2 | - | 6 - 7 | 748 | 2961181 | 66 | |
| M22632.1 | - | 6 - 7 | 704 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001158.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
gluconeogenesis
2-oxoglutarate metabolic process
oxaloacetate metabolic process
aspartate metabolic process
aspartate biosynthetic process
aspartate catabolic process
glutamate metabolic process
cellular amino acid biosynthetic process
fatty acid transport
4-hydroxyproline catabolic process
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
response to ethanol
L-kynurenine metabolic process
L-aspartate:2-oxoglutarate aminotransferase activity
phospholipid binding
kynurenine-oxoglutarate transaminase activity
amino acid binding
enzyme binding
pyridoxal phosphate binding
identical protein binding
protein homodimerization activity
poly(A) RNA binding
L-phenylalanine:2-oxoglutarate aminotransferase activity
gluconeogenesis
2-oxoglutarate metabolic process
oxaloacetate metabolic process
aspartate metabolic process
aspartate biosynthetic process
aspartate catabolic process
glutamate metabolic process
cellular amino acid biosynthetic process
fatty acid transport
4-hydroxyproline catabolic process
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
response to ethanol
L-kynurenine metabolic process
L-aspartate:2-oxoglutarate aminotransferase activity
phospholipid binding
kynurenine-oxoglutarate transaminase activity
amino acid binding
enzyme binding
pyridoxal phosphate binding
identical protein binding
protein homodimerization activity
poly(A) RNA binding
L-phenylalanine:2-oxoglutarate aminotransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016128.hg.1 | ENST00000245206.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016128.hg.1 | ENST00000425685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016128.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016128.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016128.hg.1 | TC16001158.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180802.hg.1 | TC1600010500.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000245206.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000434819.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000492378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000494627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000496461.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000564400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | ENST00000568368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_001286220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010500.hg.1 | NM_002080.3 | Refseq |

GOT2
Genomic Map