Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |