Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000492130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000492130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
ENST00000492130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024764.hg.1 |
TC06001993.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024764.hg.1 |
ENST00000193322.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024764.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202420.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202420.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202420.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202420.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202420.hg.1 |
TC0600014328.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000193322.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000467960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000472669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000477774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000492130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000492130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
ENST00000492130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014328.hg.1 |
NM_014028.3 |
Refseq |