Assay Details
| Gene Symbol: SH3D21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017002346.1 | XP_016857835.1 | 3 - 4 | 327 | 94 | |
| GenBank mRNA | AK056459.1 | - | 3 - 4 | 317 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056459.1, XM_017002346.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |

SH3D21
Genomic Map