Assay Details
| Gene Symbol: SH3D21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162530.1 | NP_001156002.1 | 2 - 3 | 192 | 139 | |
| XM_011542153.2 | XP_011540455.1 | 2 - 3 | 237 | 139 | ||
| XM_017002340.1 | XP_016857829.1 | 2 - 3 | 237 | 139 | ||
| XM_017002341.1 | XP_016857830.1 | 2 - 3 | 237 | 139 | ||
| XM_017002342.1 | XP_016857831.1 | 2 - 3 | 237 | 139 | ||
| XM_017002343.1 | XP_016857832.1 | 2 - 3 | 237 | 139 | ||
| XM_017002344.1 | XP_016857833.1 | 2 - 3 | 237 | 139 | ||
| XM_017002345.1 | XP_016857834.1 | 2 - 3 | 237 | 139 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006319.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01007305.hg.1 | TC01006319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007305.hg.1 | ENST00000426732.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007305.hg.1 | ENST00000453908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007305.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237656.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237656.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237656.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237656.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237656.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237656.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237656.hg.1 | TC0100018210.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000373139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000453908.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000496636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000505871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | ENST00000508854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_001162530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018210.hg.1 | NM_024676.4 | Refseq |

SH3D21
Genomic Map