| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007315.hg.1 |
TC01006319.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007315.hg.1 |
ENST00000312808.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007315.hg.1 |
ENST00000426732.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007315.hg.1 |
ENST00000453908.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007315.hg.1 |
ENST00000505871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007315.hg.1 |
NM_001162530.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007315.hg.1 |
NM_024676.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000373139.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000453908.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000496636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000505871.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
NM_001162530.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
NM_024676.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000373139.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000453908.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000496636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
ENST00000505871.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
NM_001162530.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
NM_024676.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100237666.hg.1 |
TC0100018210.hg.1 |
Clariom D TC |