Assay Details
| Gene Symbol: DHDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014475.3 | NP_055290.1 | 1 - 2 | 132 | 63 | |
| GenBank mRNA | AB021933.1 | - | 1 - 2 | 92 | 63 | |
| BC032730.1 | - | 1 - 2 | 143 | 5530225 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000715.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19009896.hg.1 | ENST00000221403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009896.hg.1 | ENST00000522614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009896.hg.1 | ENST00000523250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009896.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009896.hg.1 | TC19000715.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009898.hg.1 | ENST00000221403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009898.hg.1 | ENST00000522614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009898.hg.1 | ENST00000523250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009898.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009898.hg.1 | TC19000715.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009897.hg.1 | ENST00000221403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009897.hg.1 | ENST00000522614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009897.hg.1 | ENST00000523250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009897.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009897.hg.1 | TC19000715.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | TC1900008503.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165458.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | TC1900008503.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165456.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | TC1900008503.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900165457.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000221403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000520557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000522614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | ENST00000523250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008503.hg.1 | NM_014475.3 | Refseq |

DHDH
Genomic Map