Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007875.hg.1 |
ENST00000381922.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007875.hg.1 |
ENST00000546022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007875.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007875.hg.1 |
TC20000546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008144.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157195.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157195.hg.1 |
NM_015985.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157195.hg.1 |
TC2000008144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157195.hg.1 |
ENST00000381922.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157195.hg.1 |
NM_015985.3 |
Refseq |