Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020118.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020118.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020118.hg.1 |
TC12001531.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010766.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182128.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182128.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182128.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182128.hg.1 |
ENST00000309680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182128.hg.1 |
ENST00000547106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182128.hg.1 |
NM_000423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182128.hg.1 |
TC1200010766.hg.1 |
Clariom D TC |