Assay Details
Gene Symbol: FZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017412.3 | NP_059108.1 | 6 - 7 | 2081 | 92 | |
NM_145866.1 | NP_665873.1 | 5 - 6 | 2035 | 92 | ||
XM_017013841.1 | XP_016869330.1 | 4 - 5 | 1446 | 92 | ||
GenBank mRNA | AB039723.1 | - | 6 - 7 | 2032 | 92 | |
AJ272427.1 | - | 4 - 5 | 1888 | 92 | ||
AK291480.1 | - | 5 - 6 | 2030 | 92 | ||
AY005130.3 | - | 4 - 5 | 1618 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000220.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
establishment of planar polarity
neuron migration
neural tube closure
hair follicle development
positive regulation of neuroblast proliferation
G-protein coupled receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
neuron differentiation
cell proliferation in midbrain
non-canonical Wnt signaling pathway
post-anal tail morphogenesis
dopaminergic neuron axon guidance
serotonergic neuron axon guidance
inner ear morphogenesis
negative regulation of mitotic cell cycle, embryonic
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
sympathetic ganglion development
commissural neuron axon guidance
negative regulation of execution phase of apoptosis
midbrain morphogenesis
G-protein coupled receptor activity
protein binding
Wnt-protein binding
PDZ domain binding
Wnt-activated receptor activity
establishment of planar polarity
neuron migration
neural tube closure
hair follicle development
positive regulation of neuroblast proliferation
G-protein coupled receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
neuron differentiation
cell proliferation in midbrain
non-canonical Wnt signaling pathway
post-anal tail morphogenesis
dopaminergic neuron axon guidance
serotonergic neuron axon guidance
inner ear morphogenesis
negative regulation of mitotic cell cycle, embryonic
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
sympathetic ganglion development
commissural neuron axon guidance
negative regulation of execution phase of apoptosis
midbrain morphogenesis
G-protein coupled receptor activity
protein binding
Wnt-protein binding
PDZ domain binding
Wnt-activated receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000517911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000517911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003189.hg.1 | TC08000220.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08003189.hg.1 | ENST00000240093.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003189.hg.1 | ENST00000537916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003189.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003189.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003188.hg.1 | TC08000220.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08003188.hg.1 | ENST00000240093.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003188.hg.1 | ENST00000537916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003188.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003188.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149048.hg.1 | TC0800007137.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000517911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000517911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000523546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007137.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | ENST00000240093.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | ENST00000537916.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | NM_017412.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | NM_145866.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149047.hg.1 | TC0800007137.hg.1 | Clariom D TC |
