Assay Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000413.2 | NP_000404.2 | 1 - 2 | 1072 | 65 | |
| XM_011524729.1 | XP_011523031.1 | 1 - 2 | 480 | 65 | ||
| XM_011524730.1 | XP_011523032.1 | 1 - 2 | 480 | 65 | ||
| XM_011524731.1 | XP_011523033.1 | 1 - 2 | 480 | 65 | ||
| GenBank mRNA | AK127832.1 | - | 1 - 2 | 3824 | 65 | |
| AK300143.1 | - | 1 - 2 | 115 | 65 | ||
| AK300159.1 | - | 1 - 2 | 117 | 65 | ||
| AK300452.1 | - | 1 - 2 | 115 | 65 | ||
| AK316292.1 | - | 1 - 2 | 300 | 65 | ||
| BC104752.1 | - | 1 - 2 | 130 | 8143755 | 65 | |
| BC111935.1 | - | 1 - 2 | 130 | 8327403 | 65 | |
| M36263.1 | - | 1 - 2 | 113 | 65 | ||
| X13440.1 | - | 1 - 2 | 113 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000528.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
estrogen biosynthetic process
estrogen metabolic process
oxidation-reduction process
bone development
testosterone biosynthetic process
cellular response to metal ion
catalytic activity
estradiol 17-beta-dehydrogenase activity
testosterone dehydrogenase (NAD+) activity
steroid biosynthetic process
estrogen biosynthetic process
estrogen metabolic process
oxidation-reduction process
bone development
testosterone biosynthetic process
cellular response to metal ion
catalytic activity
estradiol 17-beta-dehydrogenase activity
testosterone dehydrogenase (NAD+) activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17006381.hg.1 | ENST00000225929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006381.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006381.hg.1 | TC17000528.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006384.hg.1 | ENST00000225928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006384.hg.1 | ENST00000225929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006384.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006384.hg.1 | TC17000528.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006386.hg.1 | ENST00000225928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006386.hg.1 | ENST00000225929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006386.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006386.hg.1 | TC17000528.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157520.hg.1 | TC1700007897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007896.hg.1 | ENST00000590052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007897.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157516.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157516.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157516.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157516.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157516.hg.1 | TC1700007897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000225929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000585807.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000587280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000590299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | ENST00000593215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | NM_000413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157521.hg.1 | TC1700007897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010728.hg.1 | ENST00000590513.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 157464 | - | - |

HSD17B1
Genomic Map