Assay Details
| Gene Symbol: EHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014601.3 | NP_055416.2 | 2 - 3 | 652 | 77 | |
| GenBank mRNA | AF181263.1 | - | 1 - 2 | 440 | 77 | |
| AF454952.1 | - | 1 - 2 | 457 | 77 | ||
| AK074701.1 | - | 2 - 3 | 574 | 77 | ||
| AK296907.1 | - | 3 - 4 | 544 | 77 | ||
| AK315548.1 | - | 2 - 3 | 576 | 77 | ||
| BC014445.1 | - | 2 - 3 | 563 | 4908085 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000681.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
blood coagulation
cortical actin cytoskeleton organization
endocytic recycling
protein localization to plasma membrane
membrane tubulation
positive regulation of myoblast fusion
positive regulation of endocytic recycling
nucleic acid binding
calcium ion binding
protein binding
ATP binding
GTP binding
hydrolase activity
protein domain specific binding
endocytosis
blood coagulation
cortical actin cytoskeleton organization
endocytic recycling
protein localization to plasma membrane
membrane tubulation
positive regulation of myoblast fusion
positive regulation of endocytic recycling
nucleic acid binding
calcium ion binding
protein binding
ATP binding
GTP binding
hydrolase activity
protein domain specific binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19009528.hg.1 | ENST00000263277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009528.hg.1 | ENST00000454483.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009528.hg.1 | ENST00000539439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009528.hg.1 | ENST00000540364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009528.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009528.hg.1 | TC19000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009529.hg.1 | ENST00000263277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009529.hg.1 | ENST00000454483.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009529.hg.1 | ENST00000539439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009529.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009529.hg.1 | TC19000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000596225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000596225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000596225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000602215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | ENST00000602215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009530.hg.1 | ENST00000263277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009530.hg.1 | ENST00000454483.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009530.hg.1 | ENST00000539439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009530.hg.1 | ENST00000540364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009530.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009530.hg.1 | TC19000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009525.hg.1 | ENST00000263277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009525.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009525.hg.1 | TC19000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009527.hg.1 | ENST00000263277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009527.hg.1 | ENST00000454483.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009527.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009527.hg.1 | ENST00000539439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009527.hg.1 | ENST00000540364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009527.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009527.hg.1 | TC19000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009531.hg.1 | ENST00000263277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009531.hg.1 | ENST00000454483.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009531.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009531.hg.1 | ENST00000539439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009531.hg.1 | ENST00000540364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009531.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009531.hg.1 | TC19000681.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900164797.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164797.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164797.hg.1 | TC1900008432.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164797.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164797.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164801.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164801.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164801.hg.1 | TC1900008432.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164801.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164801.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000540884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000596225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000596225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000596225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000602215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | ENST00000602215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008432.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164799.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164799.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164799.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164799.hg.1 | TC1900008432.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164799.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164799.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164799.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164802.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164802.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164802.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164802.hg.1 | TC1900008432.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164802.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164802.hg.1 | ENST00000538399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164802.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164800.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164800.hg.1 | NM_014601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164800.hg.1 | TC1900008432.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164800.hg.1 | ENST00000263277.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164800.hg.1 | NM_014601.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 108484 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 125923 | - | - |

EHD2
Genomic Map