Assay Details
| Gene Symbol: TNFRSF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001561.5 | NP_001552.2 | 4 - 5 | 468 | 77 | |
| XM_006710618.3 | XP_006710681.1 | 3 - 4 | 1115 | 77 | ||
| XM_011541386.2 | XP_011539688.1 | 3 - 4 | 483 | 77 | ||
| GenBank mRNA | BC006196.2 | - | 3 - 4 | 328 | 2924109 | 77 |
| BP381234.1 | - | 4 - 5 | 468 | 77 | ||
| L12964.2 | - | 3 - 4 | 346 | 77 | ||
| U03397.1 | - | 3 - 4 | 326 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002163.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
inflammatory response
immune response
multicellular organism development
negative regulation of cell proliferation
extrinsic apoptotic signaling pathway via death domain receptors
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of apoptotic process
protein homotrimerization
negative regulation of interleukin-10 secretion
negative regulation of interleukin-12 secretion
receptor activity
tumor necrosis factor-activated receptor activity
cytokine binding
apoptotic process
inflammatory response
immune response
multicellular organism development
negative regulation of cell proliferation
extrinsic apoptotic signaling pathway via death domain receptors
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of apoptotic process
protein homotrimerization
negative regulation of interleukin-10 secretion
negative regulation of interleukin-12 secretion
receptor activity
tumor necrosis factor-activated receptor activity
cytokine binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01033388.hg.1 | TC01002163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033388.hg.1 | ENST00000377507.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033388.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033387.hg.1 | TC01002163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033387.hg.1 | ENST00000377507.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033387.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033387.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000474475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000474475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000474475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194277.hg.1 | TC0100012734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100194278.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194278.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194278.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194278.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194278.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194278.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194278.hg.1 | TC0100012734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000377507.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000474475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000474475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000474475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000492571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | ENST00000615230.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012734.hg.1 | NM_001561.5 | Refseq |

TNFRSF9
Genomic Map