Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
TC10002942.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
ENST00000313771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
ENST00000352159.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
ENST00000355279.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
ENST00000355740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
ENST00000357339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
ENST00000371875.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
ENST00000540197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
NM_000043.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
NM_152871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
NM_152872.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
NR_028033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
NR_028034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
NR_028035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006981.hg.1 |
NR_028036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000313771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000352159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000355279.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000355740.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000357339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000479522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000484444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000487314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000488877.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000492756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000494799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000612663.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000615406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000626542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NM_000043.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NM_152871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NM_152872.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000313771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000352159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000355279.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000355740.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000357339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000479522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000484444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000487314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000488877.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000492756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000494799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000612663.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000615406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
ENST00000626542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NM_000043.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NM_152871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NM_152872.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
NR_028036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156877.hg.1 |
TC1000008388.hg.1 |
Clariom D TC |