| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008111.hg.1 |
TC10002957.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008111.hg.1 |
ENST00000370646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008111.hg.1 |
ENST00000370647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008111.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008111.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186122.hg.1 |
TC1000012489.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370642.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370646.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000370647.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
ENST00000465608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_001134670.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012489.hg.1 |
NM_138413.3 |
Refseq |