Assay Details
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003561.2 | 16 - 17 | 2982 | 148 | ||
NR_027768.1 | 14 - 15 | 3197 | 148 | |||
GenBank mRNA | AK093485.1 | - | 8 - 9 | 609 | 148 | |
AK131353.1 | - | 16 - 17 | 2975 | 148 | ||
AL834175.1 | - | 14 - 15 | 2950 | 148 | ||
BC150309.1 | - | 13 - 14 | 2589 | 148 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001717.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000312132.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000411491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000435536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000438364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000439473.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | ENST00000446373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_003561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012119.hg.1 | NR_027768.1 | Refseq |
