Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022664.hg.1 |
TC07001505.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022664.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022665.hg.1 |
TC07001505.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022665.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181762.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181762.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181762.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181762.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181762.hg.1 |
TC0700011502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000222800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000357419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000395147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000412965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000437775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000437891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000458339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000468331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000468998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000474130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000480445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000486114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_001145364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_001301058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_148912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_148913.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000222800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000357419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000395147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000412965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000437775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000437891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000458339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000468331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000468998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000474130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000480445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000486114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_001145364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_001301058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_148912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NM_148913.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181772.hg.1 |
TC0700011502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000222800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000357419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000395147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000412965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000437775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000437891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000458339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000468331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000474130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000480445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000486114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_001145364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_001301058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_148912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_148913.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000222800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000357419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000395147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000412965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000437775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000437891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000458339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000468331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000474130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000480445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000486114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_001145364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_001301058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_148912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NM_148913.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181766.hg.1 |
TC0700011502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700181761.hg.1 |
ENST00000462381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181761.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181761.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181761.hg.1 |
ENST00000462381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181761.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181761.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181761.hg.1 |
TC0700011502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000222800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000357419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000395147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000412965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000437775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000437891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000458339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000474130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000486114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_001145364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_001301058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_148912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_148913.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000222800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000357419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000395147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000412965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000437775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000437891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000458339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000474130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000486114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
ENST00000497897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_001145364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_001301058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_148912.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NM_148913.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
NR_026912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181764.hg.1 |
TC0700011502.hg.1 |
Clariom D TC |