Assay Details
| Gene Symbol: C12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031748.3 | NP_001026918.2 | 3 - 4 | 369 | 115 | |
| NM_001319247.1 | NP_001306176.1 | 3 - 4 | 369 | 115 | ||
| GenBank mRNA | AL705293.1 | - | 3 - 4 | 345 | 115 | |
| BC038754.1 | - | 3 - 4 | 366 | 5269851 | 115 | |
| BC048120.1 | - | 3 - 4 | 353 | 5266211 | 115 | |
| BI461967.1 | - | 3 - 4 | 370 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000307.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC038754.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003671.hg.1 | ENST00000324616.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003671.hg.1 | ENST00000398716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003671.hg.1 | ENST00000405531.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003671.hg.1 | ENST00000468200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003671.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003671.hg.1 | TC12000307.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151419.hg.1 | TC1200007349.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |

C12orf40
Genomic Map