Assay Details
Gene Symbol: ZNF232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320952.1 | NP_001307881.1 | 2 - 3 | 951 | 72 | |
NM_001320953.1 | NP_001307882.1 | 4 - 5 | 1238 | 72 | ||
NM_001320954.1 | NP_001307883.1 | 4 - 5 | 1183 | 72 | ||
NM_014519.3 | NP_055334.2 | 2 - 3 | 951 | 72 | ||
XM_011524006.2 | XP_011522308.1 | 3 - 4 | 617 | 72 | ||
XM_017025021.1 | XP_016880510.1 | 3 - 4 | 1168 | 72 | ||
GenBank mRNA | AK225622.1 | - | 2 - 3 | 508 | 72 | |
BC002852.1 | - | 527 | 72 | |||
BC016039.1 | - | 2 - 3 | 536 | 4635999 | 72 | |
Y15067.1 | - | 2 - 3 | 543 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000571076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000571076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000571076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000571076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000571076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000571076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009551.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000250076.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000570486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000572468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000573015.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000574735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000575538.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | ENST00000575898.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | NM_014519.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173603.hg.1 | TC1700009551.hg.1 | Clariom D TC |
