Assay Details
| Gene Symbol: KAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003884.4 | NP_003875.3 | 4 - 5 | 1113 | 101 | |
| XM_005265528.4 | XP_005265585.1 | 4 - 5 | 738 | 101 | ||
| XM_017007423.1 | XP_016862912.1 | 5 - 6 | 568 | 101 | ||
| XM_017007424.1 | XP_016862913.1 | 4 - 5 | 664 | 101 | ||
| GenBank mRNA | BC060823.1 | - | 4 - 5 | 718 | 30336561 | 101 |
| BC070075.1 | - | 4 - 5 | 783 | 30333414 | 101 | |
| U57317.2 | - | 4 - 5 | 1125 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000113.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription initiation from RNA polymerase II promoter
protein acetylation
cell cycle arrest
Notch signaling pathway
memory
negative regulation of cell proliferation
regulation of protein ADP-ribosylation
positive regulation of neuron projection development
viral process
N-terminal peptidyl-lysine acetylation
internal peptidyl-lysine acetylation
peptidyl-lysine acetylation
cellular response to insulin stimulus
cellular response to oxidative stress
positive regulation of chromatin binding
positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter
histone H3 acetylation
histone H3-K9 acetylation
negative regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of gene expression, epigenetic
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
cellular response to parathyroid hormone stimulus
positive regulation of histone H3-K14 acetylation
positive regulation of histone H3-K9 acetylation
RNA polymerase II regulatory region sequence-specific DNA binding
chromatin binding
transcription cofactor activity
transcription coactivator activity
histone acetyltransferase activity
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
transcription factor binding
acetyltransferase activity
protein kinase binding
protein complex binding
histone acetyltransferase binding
histone deacetylase binding
chromatin remodeling
transcription initiation from RNA polymerase II promoter
protein acetylation
cell cycle arrest
Notch signaling pathway
memory
negative regulation of cell proliferation
regulation of protein ADP-ribosylation
positive regulation of neuron projection development
viral process
N-terminal peptidyl-lysine acetylation
internal peptidyl-lysine acetylation
peptidyl-lysine acetylation
cellular response to insulin stimulus
cellular response to oxidative stress
positive regulation of chromatin binding
positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter
histone H3 acetylation
histone H3-K9 acetylation
negative regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of gene expression, epigenetic
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
cellular response to parathyroid hormone stimulus
positive regulation of histone H3-K14 acetylation
positive regulation of histone H3-K9 acetylation
RNA polymerase II regulatory region sequence-specific DNA binding
chromatin binding
transcription cofactor activity
transcription coactivator activity
histone acetyltransferase activity
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
transcription factor binding
acetyltransferase activity
protein kinase binding
protein complex binding
histone acetyltransferase binding
histone deacetylase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03002288.hg.1 | TC03000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002288.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002288.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002284.hg.1 | TC03000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002284.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002284.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002284.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002286.hg.1 | TC03000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002286.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002286.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000468400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147457.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147457.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147457.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147457.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147457.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147457.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147457.hg.1 | TC0300006791.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300147455.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147455.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147455.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147455.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147455.hg.1 | TC0300006791.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000468400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | ENST00000469085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006791.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147453.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147453.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147453.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147453.hg.1 | ENST00000263754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147453.hg.1 | ENST00000426228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300147453.hg.1 | NM_003884.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300147453.hg.1 | TC0300006791.hg.1 | Clariom D TC |

KAT2B
Genomic Map