Assay Details
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142318.1 | NP_001135790.1 | 1 - 2 | 116 | 113 | |
GenBank mRNA | CD517462.1 | - | 1 - 2 | 164 | 113 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001256.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001142318.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017509.hg.1 | ENST00000426362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017509.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017509.hg.1 | TC16001256.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183924.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183924.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183924.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183924.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183924.hg.1 | TC1600010809.hg.1 | Clariom D TC |
