Assay Details
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142317.1 | NP_001135789.1 | 3 - 4 | 547 | 85 | |
NM_001142318.1 | NP_001135790.1 | 3 - 4 | 445 | 85 | ||
NM_001324355.1 | NP_001311284.1 | 4 - 5 | 696 | 85 | ||
NM_017853.2 | NP_060323.1 | 3 - 4 | 596 | 85 | ||
GenBank mRNA | AK000518.1 | - | 3 - 4 | 571 | 85 | |
AK222889.1 | - | 3 - 4 | 563 | 85 | ||
AY566808.1 | - | 2 - 3 | 285 | 85 | ||
BC009646.1 | - | 3 - 4 | 537 | 3458545 | 85 | |
BM849401.1 | - | 4 - 5 | 597 | 85 | ||
CD517462.1 | - | 3 - 4 | 493 | 85 | ||
CR457240.1 | - | 2 - 3 | 285 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001256.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16017499.hg.1 | ENST00000268483.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017499.hg.1 | ENST00000423037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017499.hg.1 | ENST00000426362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017499.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017499.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017499.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017499.hg.1 | TC16001256.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008334.hg.1 | ENST00000561827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008334.hg.1 | ENST00000561827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300014091.hg.1 | ENST00000455557.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300014091.hg.1 | ENST00000455557.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300014091.hg.1 | ENST00000455557.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300014091.hg.1 | ENST00000455557.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008334.hg.1 | ENST00000561827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008334.hg.1 | ENST00000561827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183907.hg.1 | TC1600010809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183908.hg.1 | TC1600010809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000426362.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000562153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000565171.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | ENST00000569767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010809.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | ENST00000268483.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | ENST00000423037.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | NM_001142317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | NM_001142318.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | NM_017853.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183906.hg.1 | TC1600010809.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 129300 | - | - |
