Assay Details
Gene Symbol: ERICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289947.1 | NP_001276876.1 | 1 - 2 | 81 | 73 | |
GenBank mRNA | BC127676.1 | - | 1 - 2 | 81 | 40127368 | 73 |
BC150546.1 | - | 1 - 2 | 81 | 73 | ||
BQ429225.1 | - | 79 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC127676.1,NM_001289947.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169581.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169581.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169581.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169581.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169581.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169581.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169581.hg.1 | TC0200009933.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200169580.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169580.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169580.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169580.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169580.hg.1 | TC0200009933.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 167451 | - | - |
