Assay Details
Gene Symbol: ERICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289947.1 | NP_001276876.1 | 3 - 4 | 321 | 80 | |
NM_001290030.1 | NP_001276959.1 | 8 - 9 | 1108 | 80 | ||
XM_011510998.2 | XP_011509300.1 | 10 - 11 | 1240 | 80 | ||
XM_011510999.2 | XP_011509301.1 | 8 - 9 | 811 | 80 | ||
XM_011511001.2 | XP_011509303.1 | 8 - 9 | 1586 | 80 | ||
XM_017003875.1 | XP_016859364.1 | 9 - 10 | 1609 | 80 | ||
XM_017003876.1 | XP_016859365.1 | 8 - 9 | 624 | 80 | ||
GenBank mRNA | BC127676.1 | - | 3 - 4 | 321 | 40127368 | 80 |
BC150546.1 | - | 3 - 4 | 321 | 80 | ||
BQ429225.1 | - | 319 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169584.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169584.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169584.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169584.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169584.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169584.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169584.hg.1 | TC0200009933.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
