Assay Details
Gene Symbol: ALOX12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139.2 | NP_001130.1 | 5 - 6 | 912 | 58 | |
GenBank mRNA | AF038461.1 | - | 5 - 6 | 910 | 58 | |
AF059250.1 | - | 5 - 6 | 688 | 58 | ||
BC041058.1 | - | 5 - 6 | 940 | 5441563 | 58 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001100.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein lipidation
sphingolipid metabolic process
positive regulation of gene expression
arachidonic acid metabolic process
lipoxygenase pathway
positive regulation of MAPK cascade
linoleic acid metabolic process
ceramide biosynthetic process
hepoxilin biosynthetic process
oxidation-reduction process
establishment of skin barrier
positive regulation of mucus secretion
arachidonate 12-lipoxygenase activity
iron ion binding
protein binding
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
linoleate 9S-lipoxygenase activity
protein lipidation
sphingolipid metabolic process
positive regulation of gene expression
arachidonic acid metabolic process
lipoxygenase pathway
positive regulation of MAPK cascade
linoleic acid metabolic process
ceramide biosynthetic process
hepoxilin biosynthetic process
oxidation-reduction process
establishment of skin barrier
positive regulation of mucus secretion
arachidonate 12-lipoxygenase activity
iron ion binding
protein binding
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
linoleate 9S-lipoxygenase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17014794.hg.1 | ENST00000319144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17014794.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17014794.hg.1 | TC17001100.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700175174.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700175174.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700175174.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700175174.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700175174.hg.1 | TC1700009669.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000319144.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000577351.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000583276.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | ENST00000584116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009669.hg.1 | NM_001139.2 | Refseq |
