Assay Details
| Gene Symbol: SLC35B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005249157.2 | XP_005249214.1 | 1 - 2 | 262 | 53 | |
| XM_011514648.1 | XP_011512950.1 | 1 - 2 | 201 | 53 | ||
| XM_017010912.1 | XP_016866401.1 | 1 - 2 | 217 | 53 | ||
| GenBank mRNA | AK304250.1 | - | 1 - 2 | 166 | 53 | |
| BX648022.1 | - | 1 - 2 | 198 | 53 | ||
| DC357624.1 | - | 1 - 2 | 217 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK304250.1, XM_005249157.2, XM_011514648.1, XM_017010912.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
mitophagy in response to mitochondrial depolarization
xenophagy
3'-phospho-5'-adenylyl sulfate transmembrane transport
3'-phosphoadenosine 5'-phosphosulfate transmembrane transporter activity
positive regulation of defense response to virus by host
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
mitophagy in response to mitochondrial depolarization
xenophagy
3'-phospho-5'-adenylyl sulfate transmembrane transport
3'-phosphoadenosine 5'-phosphosulfate transmembrane transporter activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 143862 | - | - |

SLC35B3
Genomic Map