Assay Details
| Gene Symbol: TGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004245.3 | NP_004236.1 | 3 - 4 | 315 | 72 | |
| NM_201631.3 | NP_963925.2 | 4 - 5 | 561 | 72 | ||
| GenBank mRNA | AF035960.1 | - | 4 - 5 | 561 | 72 | |
| AF035961.1 | - | 3 - 4 | 315 | 72 | ||
| BC119009.1 | - | 4 - 5 | 561 | 40085374 | 72 | |
| BC122859.1 | - | 4 - 5 | 561 | 40085372 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001274.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011837.hg.1 | ENST00000220420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011837.hg.1 | ENST00000349114.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011837.hg.1 | ENST00000396996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011837.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011837.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011837.hg.1 | TC15001274.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167129.hg.1 | TC1500009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |

TGM5
Genomic Map