Assay Details
| Gene Symbol: POM121L10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024593.1 | 1 | 5347 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000591.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22010437.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010437.hg.1 | TC22000591.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | ENST00000441984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010436.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010436.hg.1 | TC22000591.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010438.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010438.hg.1 | TC22000591.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000416498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000416498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009192.hg.1 | ENST00000416498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000414583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000419222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | ENST00000441984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006885.hg.1 | NR_003714.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000412938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000416498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000416498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009192.hg.1 | ENST00000416498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200159009.hg.1 | TC2200008230.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200159009.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200159009.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200159010.hg.1 | TC2200008230.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200159010.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200159010.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200159008.hg.1 | TC2200008230.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200159008.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200159008.hg.1 | NR_024593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008072.hg.1 | ENST00000420508.1 | Ensembl |

POM121L10P
Genomic Map