Assay Details
| Gene Symbol: ALOX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000697.2 | NP_000688.2 | 4 - 5 | 598 | 76 | |
| GenBank mRNA | BC069557.1 | - | 4 - 5 | 576 | 7262463 | 76 |
| M35418.1 | - | 4 - 5 | 586 | 76 | ||
| M58704.1 | - | 4 - 5 | 578 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000081.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
aging
positive regulation of cell proliferation
positive regulation of endothelial cell migration
positive regulation of gene expression
negative regulation of muscle cell apoptotic process
arachidonic acid metabolic process
lipoxygenase pathway
fatty acid oxidation
positive regulation of cell growth
positive regulation of cell migration
superoxide anion generation
negative regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
linoleic acid metabolic process
positive regulation of endothelial cell differentiation
positive regulation of angiogenesis
positive regulation of cell adhesion
positive regulation of vasodilation
positive regulation of smooth muscle cell proliferation
hepoxilin metabolic process
hepoxilin biosynthetic process
positive regulation of mitochondrial depolarization
establishment of skin barrier
cellular response to lipid
reactive oxygen species metabolic process
negative regulation of platelet aggregation
leukotriene A4 metabolic process
lipoxin metabolic process
lipoxin A4 biosynthetic process
lipoxin B4 biosynthetic process
arachidonate 12-lipoxygenase activity
iron ion binding
protein binding
linoleate 13S-lipoxygenase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
hepoxilin-epoxide hydrolase activity
hepoxilin A3 synthase activity
movement of cell or subcellular component
aging
positive regulation of cell proliferation
positive regulation of endothelial cell migration
positive regulation of gene expression
negative regulation of muscle cell apoptotic process
arachidonic acid metabolic process
lipoxygenase pathway
fatty acid oxidation
positive regulation of cell growth
positive regulation of cell migration
superoxide anion generation
negative regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
linoleic acid metabolic process
positive regulation of endothelial cell differentiation
positive regulation of angiogenesis
positive regulation of cell adhesion
positive regulation of vasodilation
positive regulation of smooth muscle cell proliferation
hepoxilin metabolic process
hepoxilin biosynthetic process
positive regulation of mitochondrial depolarization
establishment of skin barrier
cellular response to lipid
reactive oxygen species metabolic process
negative regulation of platelet aggregation
leukotriene A4 metabolic process
lipoxin metabolic process
lipoxin A4 biosynthetic process
lipoxin B4 biosynthetic process
arachidonate 12-lipoxygenase activity
iron ion binding
protein binding
linoleate 13S-lipoxygenase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
hepoxilin-epoxide hydrolase activity
hepoxilin A3 synthase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000990.hg.1 | ENST00000251535.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000990.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000990.hg.1 | TC17000081.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000406228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | ENST00000480801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006717.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146377.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146377.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146377.hg.1 | ENST00000251535.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146377.hg.1 | NM_000697.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146377.hg.1 | TC1700006717.hg.1 | Clariom D TC |

ALOX12
Genomic Map